Assay Details
| Gene Symbol: Hmgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008252.3 | NP_032278.1 | 4 | 565 | 64 | |
| GenBank mRNA | AK003773.1 | - | 4 | 471 | 64 | |
| AK011467.1 | - | 4 | 472 | 64 | ||
| AK012568.1 | - | 4 | 532 | 64 | ||
| AK076075.1 | - | 4 | 501 | 64 | ||
| AK135296.1 | - | 4 | 471 | 64 | ||
| AK135297.1 | - | 4 | 471 | 64 | ||
| AK146212.1 | - | 4 | 500 | 64 | ||
| AK151164.1 | - | 4 | 479 | 64 | ||
| AK153858.1 | - | 4 | 474 | 64 | ||
| BC002050.1 | - | 4 | 445 | 3489780 | 64 | |
| BC046759.1 | - | 4 | 469 | 5693598 | 64 | |
| BC083108.1 | - | 4 | 463 | 5320702 | 64 | |
| Z46757.1 | - | 4 | 474 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of endothelial cell proliferation
base-excision repair, DNA ligation
chromatin remodeling
regulation of transcription from RNA polymerase II promoter
spermatogenesis
spermatid nucleus differentiation
male gonad development
positive regulation of nuclease activity
positive regulation of DNA binding
positive regulation of erythrocyte differentiation
positive regulation of megakaryocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to steroid hormone
positive chemotaxis
cell chemotaxis
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
DNA binding
chromatin binding
damaged DNA binding
transcription factor activity, sequence-specific DNA binding
DNA binding, bending
protein domain specific binding
chemoattractant activity
transcription regulatory region DNA binding
poly(A) RNA binding
RAGE receptor binding
positive regulation of endothelial cell proliferation
base-excision repair, DNA ligation
chromatin remodeling
regulation of transcription from RNA polymerase II promoter
spermatogenesis
spermatid nucleus differentiation
male gonad development
positive regulation of nuclease activity
positive regulation of DNA binding
positive regulation of erythrocyte differentiation
positive regulation of megakaryocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to steroid hormone
positive chemotaxis
cell chemotaxis
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
DNA binding
chromatin binding
damaged DNA binding
transcription factor activity, sequence-specific DNA binding
DNA binding, bending
protein domain specific binding
chemoattractant activity
transcription regulatory region DNA binding
poly(A) RNA binding
RAGE receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891574 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891566 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994939 | Z46757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994939 | AK146212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994939 | AK153858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994939 | BC002050.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1994939 | BC046759.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575432 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994939 | Z46757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994939 | AK146212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994939 | AK153858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994939 | BC002050.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1994939 | BC046759.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499940 | Z46757.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499940 | AK153858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499940 | BC002050.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499940 | BC046759.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P499940 | BC083108.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P485417 | Z46757.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P485417 | AK146212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P485417 | AK151164.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P485417 | AK153858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P485417 | BC002050.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891572 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25891549 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06522796 | - | - |
| Whole Mouse Genome G4122A | A_52_P485417 | Z46757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P485417 | AK146212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P485417 | AK151164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P485417 | AK153858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P485417 | BC002050.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499940 | Z46757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499940 | AK153858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499940 | BC002050.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499940 | BC046759.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P499940 | BC083108.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131111 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06522796 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06827938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06827937 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06827933 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06827942 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06827936 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06827941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891574 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891572 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891568 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25891567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25891615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07159371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06730415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07159374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06833339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06827940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06918777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06827936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06896430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06735862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06827938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25891582 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10571874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10571875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510175 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437313_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438307_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452534_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10510172 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10571870 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518350 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518352 | - | - |
| Mouse Genome 430 2.0 Array | 1438307_at | - | - |
| Mouse Genome 430 2.0 Array | 1438307_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452534_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437313_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437313_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452534_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800004233.mm.1 | NM_008252.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004233.mm.1 | NM_008252.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004233.mm.1 | TC0800000629.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800004236.mm.1 | NM_008252.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004236.mm.1 | NM_008252.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004236.mm.1 | TC0800000629.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800004240.mm.1 | NM_008252.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004240.mm.1 | NM_008252.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004240.mm.1 | TC0800000629.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800004237.mm.1 | NM_008252.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004237.mm.1 | NM_008252.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004237.mm.1 | TC0800000629.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800004241.mm.1 | NM_008252.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004241.mm.1 | NM_008252.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004241.mm.1 | TC0800000629.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800004238.mm.1 | NM_008252.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004238.mm.1 | NM_008252.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004238.mm.1 | TC0800000629.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800004239.mm.1 | NM_008252.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004239.mm.1 | NM_008252.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004239.mm.1 | TC0800000629.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437313_PM_x_at | - | - |
| Mouse Expression Set 430 | 1452534_a_at | - | - |
| Mouse Expression Set 430 | 1437313_x_at | - | - |
| Mouse Expression Set 430 | 1452534_PM_a_at | - | - |
| Murine Genome U74v2 Set | 93250_r_at | - | - |
| Murine Genome U74v2 Set | 93250_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452534_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437313_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437313_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452534_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452534_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452534_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437313_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437313_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 797171 | - | - |
| ABI Mouse Genome Survey Microarray | 903048 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2640603 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2640603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002640603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2749954 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2749954 | - | - |
| mouse-6 v1.1 (Illumina) | scl097165.1_204-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2640603 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl097165.1_204-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2640603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3156604 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2749954 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3156604 | - | - |

Hmgb2
Genomic Map