Assay Details
Gene Symbol: Gm6086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039219.2 | NP_001034308.1 | 8 | 3957 | 109 | |
GenBank mRNA | AK145447.1 | - | 8 | 3958 | 109 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969406 | NM_001039219.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969406 | AK145447.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499538 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873752 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545669 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40975 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164072 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102127 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152653 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969406 | NM_001039219.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969406 | AK145447.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09561596 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152653 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00136910 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00136910 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20457732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P32978298 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P32978302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P32978296 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10348905 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348907 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348904 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348909 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348908 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348903 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348901 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348898 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348900 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10348899 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10348896 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10348906 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10348902 | - | - |
Affymetrix Mouse Clariom S Array | PSR0100008111.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008111.mm.1 | TC0100000951.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008105.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008105.mm.1 | TC0100000951.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008112.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008112.mm.1 | TC0100000951.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008106.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008106.mm.1 | TC0100000951.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008107.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008107.mm.1 | TC0100000951.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008108.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008108.mm.1 | TC0100000951.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008109.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008109.mm.1 | TC0100000951.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008110.mm.1 | NM_001039219.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008110.mm.1 | TC0100000951.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1228902 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3087413 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3015989 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3015989 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3087413 | - | - |
