Assay Details
Gene Symbol: Zfp488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013777.2 | NP_001013799.2 | 2 | 1127 | 100 | |
GenBank mRNA | BC089025.1 | - | 2 | 1127 | 30618054 | 100 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
oligodendrocyte development
negative regulation of transcription, DNA-templated
positive regulation of oligodendrocyte differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
metal ion binding
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
oligodendrocyte development
negative regulation of transcription, DNA-templated
positive regulation of oligodendrocyte differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29527233 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29527232 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29527230 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093924 | NM_001013777.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2093924 | BC089025.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980432 | AK035047.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980432 | NM_001013777.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980432 | BC089025.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333100 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588827 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320429 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410927 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599079 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57292 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143896 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400161 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83059 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30842 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70010 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980432 | AK035047.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980432 | NM_001013777.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980432 | BC089025.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093924 | NM_001013777.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2093924 | BC089025.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527216 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527243 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527237 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527213 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527244 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527234 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527219 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527235 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527239 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527240 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527238 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527241 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527221 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527218 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527233 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527215 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29527223 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02830059 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02830065 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02830048 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02830048 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02830065 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29527230 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29527232 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29527233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29527237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11190374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527205 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527212 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527235 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527244 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11146510 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11213164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11798360 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11798361 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11558423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527233 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11558406 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527229 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11558398 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527220 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11558408 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11672506 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29527186 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10418897 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10418896 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10418895 | - | - |
Affymetrix Mouse Clariom S Array | PSR1400013366.mm.1 | NM_001013777.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400013366.mm.1 | NM_001013777.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400013366.mm.1 | TC1400001715.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400013367.mm.1 | NM_001013777.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400013367.mm.1 | NM_001013777.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400013367.mm.1 | TC1400001715.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2536086 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2808383 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2808383 | - | - |
