Assay Details
Gene Symbol: C330021F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024728.2 | NP_001019899.1 | 6 | 1535 | 133 | |
GenBank mRNA | AK049309.1 | - | 6 | 1538 | 133 | |
AK166699.1 | - | 2 | 1050 | 133 | ||
BC089480.1 | - | 4 | 1298 | 30307882 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200002581.rn
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077555 | BC089480.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077555 | AK049309.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077555 | AK166699.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077555 | NM_001024728.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578556 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729654 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773539 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866502 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447350 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825200 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780132 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818797 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150500 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22120 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177420 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108203 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179898 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077555 | BC089480.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077555 | AK049309.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077555 | AK166699.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077555 | NM_001024728.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P121361 | AK039607.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25614606 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25614607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25614610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25614634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29215844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29215847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29215843 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179898 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03810542 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108203 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177420 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134026 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108203 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179898 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614605 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614543 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614575 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25614620 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614593 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727171 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614564 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614603 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614534 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614539 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06692011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06797675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614589 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614541 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727174 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06692017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614595 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06993418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727183 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614609 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614636 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614602 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727199 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07404529 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614610 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614561 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25614605 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25727160 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10569826 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569827 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569829 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569825 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569828 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10569824 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10576727 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10569823 | - | - |
Affymetrix Mouse Clariom S Array | PSR0800000173.mm.1 | NM_001024728.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000173.mm.1 | NM_001024728.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000173.mm.1 | TC0800000012.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800000170.mm.1 | NM_001024728.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000170.mm.1 | NM_001024728.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000170.mm.1 | TC0800000012.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800000162.mm.1 | NM_001024728.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000162.mm.1 | NM_001024728.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000162.mm.1 | TC0800000012.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800000171.mm.1 | NM_001024728.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000171.mm.1 | NM_001024728.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000171.mm.1 | TC0800000012.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800000163.mm.1 | NM_001024728.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000163.mm.1 | NM_001024728.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000163.mm.1 | TC0800000012.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800000166.mm.1 | NM_001024728.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000166.mm.1 | NM_001024728.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000166.mm.1 | TC0800000012.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800000167.mm.1 | NM_001024728.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000167.mm.1 | NM_001024728.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800000167.mm.1 | TC0800000012.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 561061 | - | - |
ABI Mouse Genome Survey Microarray | 933113 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102940053 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225930 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1225930 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243111 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1259776 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102940053 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102940053 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3102795 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3102795 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243111 | - | - |
