Assay Details
| Gene Symbol: Serpina3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033335.3 | NP_001028507.2 | 6 | 1532 | 95 | |
| NM_001168294.1 | NP_001161766.1 | 6 | 1573 | 95 | ||
| NM_001168295.1 | NP_001161767.1 | 5 | 1361 | 95 | ||
| GenBank mRNA | AK138954.1 | - | 6 | 1533 | 95 | |
| BC049975.1 | - | 5 | 1351 | 95 | ||
| BC139132.1 | - | 5 | 1421 | 8862154 | 95 | |
| BC139133.1 | - | 5 | 1421 | 8862155 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104975 | BC139133.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104975 | BC139132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104975 | NM_001168294.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104975 | NM_001168295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2104975 | AK138954.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155960 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142226 | NR_033450.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104975 | BC139133.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104975 | BC139132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104975 | NM_001168294.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104975 | NM_001168295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2104975 | AK138954.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28702048 | - | - |
| Whole Mouse Genome G4122A | A_52_P419678 | BC139133.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419678 | BC139132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419678 | NM_001168294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419678 | NM_001168295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419678 | AK138954.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419679 | BC139133.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419679 | BC139132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419679 | NM_001168294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419679 | NM_001168295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P419679 | AK138954.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28702064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10337141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10251317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10127200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28702077 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398042 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398041 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398044 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398043 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398039 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007408.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007408.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007408.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007408.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007408.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007407.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007407.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007407.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007407.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007407.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007412.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007412.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007412.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007412.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007412.mm.1 | NM_001168295.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007412.mm.1 | NM_001168295.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007412.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200018815.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200018815.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200018815.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200018815.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200018815.mm.1 | NM_001168295.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200018815.mm.1 | NM_001168295.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200018815.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007405.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007405.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007405.mm.1 | NM_001168295.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007405.mm.1 | NM_001168295.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007405.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007413.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007413.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007413.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007413.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007413.mm.1 | NM_001168295.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007413.mm.1 | NM_001168295.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007413.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007414.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007414.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007414.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007414.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007414.mm.1 | NM_001168295.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007414.mm.1 | NM_001168295.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007414.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007415.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007415.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007415.mm.1 | NM_001168294.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007415.mm.1 | NM_001168294.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007415.mm.1 | NM_001168295.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007415.mm.1 | NM_001168295.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007415.mm.1 | TC1200002524.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007406.mm.1 | NM_001033335.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007406.mm.1 | NM_001033335.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007406.mm.1 | TC1200002524.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2742861 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2742861 | - | - |

Serpina3f
Genomic Map