Assay Details
| Gene Symbol: Gm5627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033301.1 | 2 | 339 | 62 | ||
| GenBank mRNA | AK041874.1 | - | 2 | 342 | 62 | |
| BC147467.1 | - | 2 | 93 | 9056109 | 62 | |
| BC147470.1 | - | 2 | 93 | 9056112 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm5627
Genomic Map