Assay Details
| Gene Symbol: Gm4787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 2259 | 117 | |
| GenBank mRNA | BC141094.1 | - | 1 | 2237 | 9055706 | 117 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm4787
Genomic Map