Assay Details
| Gene Symbol: Nufip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024205.2 | NP_001019376.1 | 1 - 2 | 367 | 75 | |
| GenBank mRNA | AK030113.1 | - | 1 - 2 | 368 | 75 | |
| AK122494.1 | - | 1 - 2 | 351 | 75 | ||
| AK132603.1 | - | 335 | 75 | |||
| AK142954.1 | - | 1 - 2 | 338 | 75 | ||
| AK152336.1 | - | 1 - 2 | 338 | 75 | ||
| AK160601.1 | - | 1 - 2 | 368 | 75 | ||
| AK170764.1 | - | 1 - 2 | 365 | 75 | ||
More Information
Set Membership: |
|

Nufip2
Genomic Map