Assay Details
Gene Symbol: Ccdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028804.1 | NP_083080.1 | 1 | 342 | 93 | |
GenBank mRNA | AK009833.1 | - | 1 | 186 | 93 | |
BC058616.1 | - | 1 | 342 | 5686139 | 93 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
