Assay Details
| Gene Symbol: Tfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291765.1 | NP_001278694.1 | 4 - 5 | 305 | 96 | |
| NM_001291766.1 | NP_001278695.1 | 3 - 4 | 299 | 96 | ||
| NM_001291768.1 | NP_001278697.1 | 3 - 4 | 238 | 96 | ||
| NM_009361.3 | NP_033387.1 | 4 - 5 | 366 | 96 | ||
| GenBank mRNA | AK013180.1 | - | 3 - 4 | 268 | 96 | |
| AK019997.1 | - | 4 - 5 | 306 | 96 | ||
| AK030820.1 | - | 4 - 5 | 367 | 96 | ||
| AK154037.1 | - | 3 - 4 | 206 | 96 | ||
| BC058633.1 | - | 4 - 5 | 349 | 96 | ||
| BC132567.1 | - | 3 - 4 | 211 | 40130844 | 96 | |
| BC132569.1 | - | 3 - 4 | 211 | 40130846 | 96 | |
| BC145344.1 | - | 3 - 4 | 211 | 9053900 | 96 | |
| BY089512.1 | - | 3 - 4 | 215 | 96 | ||
| X72310.1 | - | 3 - 4 | 240 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK013180.1, AK154037.1, NM_001291766.1, NM_001291768.1 transcripts.
Set Membership: |
|

Tfdp1
Genomic Map