Assay Details
| Gene Symbol: Tfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291765.1 | NP_001278694.1 | 6 - 7 | 591 | 65 | |
| NM_001291766.1 | NP_001278695.1 | 5 - 6 | 585 | 65 | ||
| NM_001291768.1 | NP_001278697.1 | 5 - 6 | 524 | 65 | ||
| NM_009361.3 | NP_033387.1 | 6 - 7 | 652 | 65 | ||
| GenBank mRNA | AK013180.1 | - | 5 - 6 | 554 | 65 | |
| AK019997.1 | - | 6 - 7 | 592 | 65 | ||
| AK030820.1 | - | 6 - 7 | 653 | 65 | ||
| AK154037.1 | - | 5 - 6 | 492 | 65 | ||
| BC132567.1 | - | 5 - 6 | 497 | 40130844 | 65 | |
| BC132569.1 | - | 5 - 6 | 497 | 40130846 | 65 | |
| BC145344.1 | - | 5 - 6 | 497 | 9053900 | 65 | |
| X72310.1 | - | 5 - 6 | 526 | 65 | ||
More Information
Set Membership: |
|

Tfdp1
Genomic Map