Assay Details
| Gene Symbol: Tfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291765.1 | NP_001278694.1 | 11 - 12 | 1204 | 67 | |
| NM_001291766.1 | NP_001278695.1 | 10 - 11 | 1198 | 67 | ||
| NM_001291768.1 | NP_001278697.1 | 10 - 11 | 1137 | 67 | ||
| NM_009361.3 | NP_033387.1 | 11 - 12 | 1265 | 67 | ||
| GenBank mRNA | AK013180.1 | - | 10 - 11 | 1167 | 67 | |
| AK030820.1 | - | 11 - 12 | 1266 | 67 | ||
| AK154037.1 | - | 10 - 11 | 1105 | 67 | ||
| BC058633.1 | - | 10 - 11 | 1480 | 67 | ||
| BC132567.1 | - | 10 - 11 | 1110 | 40130844 | 67 | |
| BC132569.1 | - | 10 - 11 | 1110 | 40130846 | 67 | |
| BC145344.1 | - | 10 - 11 | 1110 | 9053900 | 67 | |
| X72310.1 | - | 10 - 11 | 1139 | 67 | ||
More Information
Set Membership: |
|

Tfdp1
Genomic Map