Assay Details
| Gene Symbol: Tpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009429.3 | NP_033455.1 | 3 - 4 | 470 | 150 | |
| GenBank mRNA | AK149695.1 | - | 3 - 4 | 396 | 150 | |
| AK150300.1 | - | 3 - 4 | 397 | 150 | ||
| AK150500.1 | - | 3 - 4 | 396 | 150 | ||
| AK150858.1 | - | 3 - 4 | 397 | 150 | ||
| AK151059.1 | - | 3 - 4 | 398 | 150 | ||
| AK151266.1 | - | 3 - 4 | 396 | 150 | ||
| AK151375.1 | - | 3 - 4 | 396 | 150 | ||
| AK151894.1 | - | 3 - 4 | 396 | 150 | ||
| AK152690.1 | - | 3 - 4 | 396 | 150 | ||
| AK152808.1 | - | 3 - 4 | 397 | 150 | ||
| AK159516.1 | - | 3 - 4 | 396 | 150 | ||
| AK159565.1 | - | 3 - 4 | 396 | 150 | ||
| AK165736.1 | - | 3 - 4 | 396 | 150 | ||
| AK166584.1 | - | 3 - 4 | 399 | 150 | ||
| AK167413.1 | - | 3 - 4 | 396 | 150 | ||
| AK167936.1 | - | 3 - 4 | 396 | 150 | ||
| AK169730.1 | - | 3 - 4 | 398 | 150 | ||
| BC092381.1 | - | 3 - 4 | 356 | 6824238 | 150 | |
| X06407.1 | - | 3 - 4 | 397 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730266 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730264 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730269 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730262 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164221 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145879 | AA762583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145879 | NR_002218.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145879 | X06407.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145879 | AK166584.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2145879 | AK167413.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075013 | AK203479.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075013 | AA762583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075013 | NR_002218.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075013 | X06407.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2075013 | AK166584.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243831 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104798 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075013 | AK203479.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075013 | AA762583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075013 | NR_002218.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075013 | X06407.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2075013 | AK166584.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145879 | AA762583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145879 | NR_002218.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145879 | X06407.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145879 | AK166584.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2145879 | AK167413.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29730266 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104798 | - | - |
| Whole Mouse Genome G4122A | A_52_P667098 | AK203479.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P667098 | AU035702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P667098 | NR_002218.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P667098 | X06407.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P667098 | AK166584.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119746 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104798 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11342234 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11342240 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11342238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11342230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11342235 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11342231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11342236 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730264 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730266 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730261 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730260 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29730269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29730258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730285 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11621984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11724217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11621980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11347683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11685904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11342234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11342231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11732851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11419912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11211538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29730254 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10416499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416500 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416502 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416501 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10584351 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416643_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416642_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10416496 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601264 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10546702 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10584350 | - | - |
| Mouse Genome 430 2.0 Array | 1416643_at | - | - |
| Mouse Genome 430 2.0 Array | 1416643_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400008526.mm.1 | NM_009429.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008526.mm.1 | NM_009429.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008526.mm.1 | TC1400001061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400008519.mm.1 | NM_009429.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008519.mm.1 | NM_009429.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008519.mm.1 | TC1400001061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400008521.mm.1 | NM_009429.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008521.mm.1 | NM_009429.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008521.mm.1 | TC1400001061.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400008527.mm.1 | NM_009429.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008527.mm.1 | NM_009429.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008527.mm.1 | TC1400001061.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416643_at | - | - |
| Mouse Expression Set 430 | 1416642_a_at | - | - |
| Mouse Expression Set 430 | 1416643_PM_at | - | - |
| Mouse Expression Set 430 | 1416642_PM_a_at | - | - |
| Murine Genome U74v2 Set | 102599_PM_at | - | - |
| Murine Genome U74v2 Set | 102599_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416642_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416642_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416643_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416643_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 894873 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102570132 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102570132 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3450035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003450035 | - | - |
| mouse-6 v1.1 (Illumina) | scl0022070.1_31-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_6678436-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3450035 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102570132 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022070.1_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102570132 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3450035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2858106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2858106 | - | - |

Tpt1
Genomic Map