Assay Details
| Gene Symbol: Vegfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025250.3 | NP_001020421.2 | 7 - 8 | 1651 | 83 | |
| NM_001110266.1 | NP_001103736.1 | 4 - 5 | 961 | 83 | ||
| NM_001110267.1 | NP_001103737.1 | 3 - 4 | 889 | 83 | ||
| NM_001287056.1 | NP_001273985.1 | 7 - 8 | 1651 | 83 | ||
| NM_001287057.1 | NP_001273986.1 | 6 - 7 | 1579 | 83 | ||
| NM_009505.4 | NP_033531.3 | 6 - 7 | 1579 | 83 | ||
| GenBank mRNA | AW913188.1 | - | 2 - 3 | 209 | 83 | |
| BC061468.1 | - | 6 - 7 | 952 | 6816435 | 83 | |
More Information
Set Membership: |
|

Vegfa
Genomic Map