Assay Details
| Gene Symbol: Adra1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271760.1 | NP_001258689.1 | 3 - 4 | 1914 | 64 | |
| NM_013461.4 | NP_038489.2 | 3 - 4 | 2377 | 64 | ||
| GenBank mRNA | AK079597.1 | - | 3 - 4 | 2370 | 64 | |
| BC113139.1 | - | 3 - 4 | 1609 | 40048262 | 64 | |
More Information
Set Membership: |
|

Adra1a
Genomic Map