Assay Details
| Gene Symbol: Cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013494.3 | NP_038522.2 | 4 - 5 | 880 | 72 | |
| GenBank mRNA | AK002935.1 | - | 4 - 5 | 525 | 72 | |
| AK013509.1 | - | 4 - 5 | 577 | 72 | ||
| AK047891.1 | - | 4 - 5 | 871 | 72 | ||
| AK160097.1 | - | 4 - 5 | 895 | 72 | ||
| BC010197.1 | - | 4 - 5 | 880 | 3157558 | 72 | |
| U23184.1 | - | 4 - 5 | 789 | 72 | ||
| X61232.1 | - | 4 - 5 | 828 | 72 | ||
More Information
Set Membership: |
|

Cpe
Genomic Map