Assay Details
| Gene Symbol: Ndufa12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025551.3 | NP_079827.2 | 2 - 3 | 206 | 130 | |
| GenBank mRNA | AK003131.1 | - | 2 - 3 | 179 | 130 | |
| AK003322.1 | - | 2 - 3 | 179 | 130 | ||
| AK008876.1 | - | 2 - 3 | 179 | 130 | ||
| AK010460.1 | - | 2 - 3 | 207 | 130 | ||
| AK032774.1 | - | 2 - 3 | 180 | 130 | ||
| AK134765.1 | - | 2 - 3 | 179 | 130 | ||
| AK153240.1 | - | 2 - 3 | 179 | 130 | ||
| AK167943.1 | - | 2 - 3 | 172 | 130 | ||
| BC004633.1 | - | 1636 | 130 | |||
| BC086922.1 | - | 2 - 3 | 211 | 6751147 | 130 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufa12
Genomic Map