Assay Details
| Gene Symbol: Tmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290751.1 | NP_001277680.1 | 5 - 6 | 632 | 106 | |
| NM_025868.4 | NP_080144.1 | 7 - 8 | 932 | 106 | ||
| GenBank mRNA | AK009759.1 | - | 6 - 7 | 620 | 106 | |
| AK075750.1 | - | 7 - 8 | 934 | 106 | ||
| AK145245.1 | - | 6 - 7 | 641 | 106 | ||
| AK170587.1 | - | 6 - 7 | 629 | 106 | ||
| BC013544.1 | - | 6 - 7 | 614 | 4242418 | 106 | |
| BC110994.1 | - | 6 - 7 | 615 | 5349078 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052495 | BC110994.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052495 | AK145245.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052495 | AK170587.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052495 | BC013544.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052495 | NM_025868.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2023777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79853 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052495 | BC110994.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052495 | AK145245.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052495 | AK170587.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052495 | BC013544.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052495 | NM_025868.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2023777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393956 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04591140 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04591122 | - | - |
| Whole Mouse Genome G4122A | A_52_P537492 | BC110994.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537492 | AK145245.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537492 | AK170587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537492 | BC013544.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P537492 | NM_025868.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P617080 | BC110994.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P617080 | AK145245.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P617080 | AK170587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P617080 | BC013544.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P617080 | NM_025868.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135477 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04591122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01279406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01279412 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01279404 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01279397 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01279417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01279413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01696180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01952926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21393950 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10484433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484434 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447355 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484439 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10484435 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437454_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452130_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10484431 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10447354 | - | - |
| Mouse Genome 430 2.0 Array | 1437454_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437454_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452130_at | - | - |
| Mouse Genome 430 2.0 Array | 1452130_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200033508.mm.1 | NM_025868.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200033508.mm.1 | TC0200003873.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437454_a_at | - | - |
| Mouse Expression Set 430 | 1452130_PM_at | - | - |
| Mouse Expression Set 430 | 1437454_PM_a_at | - | - |
| Mouse Expression Set 430 | 1452130_at | - | - |
| Murine Genome U74v2 Set | 96649_at | - | - |
| Murine Genome U74v2 Set | 96649_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437454_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452130_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452130_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437454_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 321856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6400725 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2598532 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755355 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2598532 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755355 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003075.1_15-S | - | - |
| mouse-6 v1.1 (Illumina) | scl066958.2_91-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840286 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400725 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066958.2_91-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003075.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840286 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400725 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2598532 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755355 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066958.2_91-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003075.1_15-S | - | - |

Tmx2
Genomic Map