Assay Details
| Gene Symbol: Rpl22l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026517.2 | NP_080793.1 | 3 - 4 | 240 | 145 | |
| GenBank mRNA | AK008915.1 | - | 3 - 4 | 242 | 145 | |
| AK010265.1 | - | 3 - 4 | 220 | 145 | ||
| AK013966.1 | - | 3 - 4 | 240 | 145 | ||
| BC026533.1 | - | 3 - 4 | 220 | 5030430 | 145 | |
| BC120716.1 | - | 3 - 4 | 315 | 40129639 | 145 | |
| BC120718.1 | - | 3 - 4 | 315 | 40129641 | 145 | |
| Gene Symbol: Gm15421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004442.1 | 1 | 557 | 145 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019228 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019224 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019226 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019227 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107357 | NR_004442.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159026 | BC026533.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159026 | NM_026517.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159026 | AK008915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159026 | AK013966.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159026 | AK170359.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352903 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119800 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107357 | NR_004442.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159026 | BC026533.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159026 | NM_026517.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159026 | AK008915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159026 | AK013966.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159026 | AK170359.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P218814 | BC026533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P218814 | BC120716.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P218814 | BC120718.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P218814 | NM_026517.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P218814 | AK008915.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019201 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22019225 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119800 | - | - |
| Whole Mouse Genome G4122A | A_52_P65897 | AK170359.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218814 | BC026533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218814 | BC120716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218814 | BC120718.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218814 | NM_026517.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P218814 | AK008915.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101340 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121693 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119800 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02867358 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02867370 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02867339 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02867344 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019224 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019225 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019227 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019228 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22019226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22019192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019203 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03136534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019205 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02867354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019217 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019240 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019200 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03136533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02733363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02867231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02867342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22019255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02867370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02416105 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10491195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491194 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528475 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417126_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10491191 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528474 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528474 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10491191 | - | - |
| Mouse Genome 430 2.0 Array | 1417126_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417126_PM_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300000974.mm.1 | NM_026517.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000974.mm.1 | NM_026517.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000974.mm.1 | TC0300000182.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018559.mm.1 | TC0500002037.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018559.mm.1 | NR_004442.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000969.mm.1 | TC0300000182.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000976.mm.1 | NM_026517.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000976.mm.1 | NM_026517.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000976.mm.1 | TC0300000182.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000973.mm.1 | NM_026517.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000973.mm.1 | NM_026517.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000973.mm.1 | TC0300000182.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300000975.mm.1 | NM_026517.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000975.mm.1 | NM_026517.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300000975.mm.1 | TC0300000182.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417126_a_at | - | - |
| Mouse Expression Set 430 | 1417126_PM_a_at | - | - |
| Murine Genome U74v2 Set | 93987_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93987_f_at | - | - |
| Murine Genome U74v2 Set | 93987_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93987_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417126_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417126_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417126_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417126_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 590982 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5720112 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2678755 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2678755 | - | - |
| mouse-6 v1.1 (Illumina) | scl068028.3_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720112 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl068028.3_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2678755 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2678755 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068028.3_20-S | - | - |

Rpl22l1
Genomic Map