Assay Details
| Gene Symbol: Fdps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253751.1 | NP_001240680.1 | 9 - 10 | 1075 | 85 | |
| NM_134469.4 | NP_608219.1 | 8 - 9 | 859 | 85 | ||
| GenBank mRNA | AF309508.1 | - | 7 - 8 | 764 | 85 | |
| AK088601.1 | - | 8 - 9 | 779 | 85 | ||
| AK140881.1 | - | 8 - 9 | 851 | 85 | ||
| AK151742.1 | - | 8 - 9 | 788 | 85 | ||
| AK166026.1 | - | 8 - 9 | 778 | 85 | ||
| AK168008.1 | - | 8 - 9 | 778 | 85 | ||
| BC048497.1 | - | 8 - 9 | 837 | 6531586 | 85 | |
| BC087886.1 | - | 8 - 9 | 782 | 6595030 | 85 | |
| CT010365.1 | - | 7 - 8 | 731 | 85 | ||
More Information
Set Membership: |
|

Fdps
Genomic Map