Assay Details
| Gene Symbol: Tufm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163713.1 | NP_001157185.1 | 2 - 3 | 337 | 90 | |
| NM_172745.3 | NP_766333.1 | 2 - 3 | 337 | 90 | ||
| GenBank mRNA | AK075857.1 | - | 2 - 3 | 261 | 90 | |
| AK084724.1 | - | 2 - 3 | 318 | 90 | ||
| AK152858.1 | - | 2 - 3 | 332 | 90 | ||
| AK153135.1 | - | 2 - 3 | 332 | 90 | ||
| BC060959.1 | - | 2 - 3 | 314 | 30379757 | 90 | |
| BC100596.1 | - | 2 - 3 | 330 | 6762419 | 90 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tufm
Genomic Map