Assay Details
| Gene Symbol: Olfr670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207146.1 | NP_997029.1 | 1 | 628 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981604 | NM_207146.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981604 | NM_207146.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P524345 | NM_207146.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25390121 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01595605 | - | - |
| Whole Mouse Genome G4122A | A_52_P524345 | NM_207146.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06204310 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06323266 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06156333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06156330 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06323269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25390134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06076745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06323263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06204310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06156332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06381420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25390100 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566410 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566409 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700033041.mm.1 | NM_207146.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033041.mm.1 | NM_207146.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700033041.mm.1 | TC0700003961.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 385313 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2540726 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2997653 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2997653 | - | - |

Olfr670
Genomic Map