Assay Details
| Gene Symbol: Methig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024672.3 | NP_001019843.1 | 1 - 2 | 508 | 92 | |
| GenBank mRNA | AB099516.1 | - | 1 - 2 | 545 | 92 | |
| AK144032.1 | - | 1 - 2 | 509 | 92 | ||
| BC139180.1 | - | 1 - 2 | 542 | 8862202 | 92 | |
| BC139182.1 | - | 1 - 2 | 542 | 8862204 | 92 | |
| Gene Symbol: Higd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB110950.1 | - | 1 - 2 | 559 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047330 | AK144032.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047330 | NM_001024672.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3245 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 849650 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 775805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10582 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136975 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109190 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144641 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047330 | AK144032.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047330 | NM_001024672.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485287 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30485294 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109190 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08481305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08481300 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08481315 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08481324 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136975 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144641 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144641 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136975 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109190 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12074051 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12074053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30485471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10389651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10170224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10389648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10389660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10389647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28349790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10389644 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10426895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426903 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426901 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10426902 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10426898 | - | - |
| Mouse Genome 430 2.0 Array | 1447356_at | - | - |
| Mouse Genome 430 2.0 Array | 1447356_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421184_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1421184_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 554292_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 554292_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500008267.mm.1 | NM_001002900.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008267.mm.1 | NM_001002900.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008267.mm.1 | NM_001024672.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008267.mm.1 | NM_001024672.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008267.mm.1 | TC1500001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008270.mm.1 | NM_001002900.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008270.mm.1 | NM_001002900.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008270.mm.1 | NM_001024672.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008270.mm.1 | NM_001024672.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008270.mm.1 | TC1500001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008268.mm.1 | NM_001002900.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008268.mm.1 | NM_001002900.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008268.mm.1 | NM_001024672.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008268.mm.1 | NM_001024672.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008268.mm.1 | TC1500001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008273.mm.1 | NM_001024672.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008273.mm.1 | NM_001024672.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008273.mm.1 | TC1500001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008256.mm.1 | NM_001024672.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008256.mm.1 | NM_001024672.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008256.mm.1 | NM_199477.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008256.mm.1 | NM_199477.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008256.mm.1 | TC1500001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008266.mm.1 | NM_001002900.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008266.mm.1 | NM_001002900.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008266.mm.1 | NM_001024672.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008266.mm.1 | NM_001024672.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008266.mm.1 | TC1500001044.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008255.mm.1 | TC1500001044.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421184_PM_a_at | - | - |
| Mouse Expression Set 430 | 1421184_a_at | - | - |
| Mouse Expression Set 430 | 1447356_at | - | - |
| Mouse Expression Set 430 | 1447356_PM_at | - | - |
| Murine Genome U74v2 Set | 131101_at | - | - |
| Murine Genome U74v2 Set | 131101_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421184_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421184_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 825000 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2941575 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2941575 | - | - |

Methig1
Genomic Map