Assay Details
| Gene Symbol: Vegfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025250.3 | NP_001020421.2 | 5 - 6 | 1447 | 87 | |
| NM_001110266.1 | NP_001103736.1 | 2 - 3 | 757 | 87 | ||
| NM_001287056.1 | NP_001273985.1 | 5 - 6 | 1447 | 87 | ||
| GenBank mRNA | AY120866.1 | - | 3 - 4 | 325 | 87 | |
| BQ554097.1 | - | 2 - 3 | 402 | 87 | ||
More Information
Set Membership: |
|

Vegfa
Genomic Map