Assay Details
| Gene Symbol: Vegfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025257.3 | NP_001020428.2 | 5 - 6 | 1442 | 87 | |
| NM_001110268.1 | NP_001103738.1 | 2 - 3 | 752 | 87 | ||
| NM_001287058.1 | NP_001273987.1 | 5 - 6 | 1442 | 87 | ||
| GenBank mRNA | BQ832724.1 | - | 4 - 5 | 343 | 87 | |
More Information
Set Membership: |
|

Vegfa
Genomic Map