Assay Details
| Gene Symbol: Rpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048057.1 | NP_001041522.1 | 3 - 4 | 184 | 71 | |
| NM_001048058.1 | NP_001041523.1 | 2 - 3 | 110 | 71 | ||
| NM_023372.2 | NP_075861.1 | 3 - 4 | 167 | 71 | ||
| GenBank mRNA | AB037665.1 | - | 3 - 4 | 186 | 71 | |
| AK002659.1 | - | 3 - 4 | 134 | 71 | ||
| AK010277.1 | - | 3 - 4 | 121 | 71 | ||
| AK010518.2 | - | 3 - 4 | 120 | 71 | ||
| AK012391.1 | - | 3 - 4 | 120 | 71 | ||
| AK151517.1 | - | 3 - 4 | 121 | 71 | ||
| BC034339.1 | - | 3 - 4 | 123 | 3985781 | 71 | |
| BC047046.1 | - | 2 - 3 | 110 | 71 | ||
| BC055346.1 | - | 3 - 4 | 119 | 5003663 | 71 | |
| BC081447.1 | - | 2 - 3 | 84 | 71 | ||
| BC094258.1 | - | 2 - 3 | 115 | 1245753 | 71 | |
| BC094644.1 | - | 2 - 3 | 116 | 3328874 | 71 | |
| BU921494.1 | - | 3 - 4 | 167 | 71 | ||
| CJ042718.1 | - | 3 - 4 | 71 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149045 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149047 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955059 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091308 | BU921494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091308 | BC094644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091308 | NM_001048057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091308 | NM_001048058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091308 | NM_023372.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23150 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138024 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091308 | BU921494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091308 | BC094644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091308 | NM_001048057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091308 | NM_001048058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091308 | NM_023372.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955059 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227332 | BU921494.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227332 | BC094644.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227332 | NM_001048057.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227332 | NM_001048058.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P227332 | NM_023372.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28149070 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138024 | - | - |
| Whole Mouse Genome G4122A | A_51_P227332 | BU921494.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227332 | BC094644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227332 | NM_001048057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227332 | NM_001048058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P227332 | NM_023372.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127840 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138024 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09428264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09428266 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09135146 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149048 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149047 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28149045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28148999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28149014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149062 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09325190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09010217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09482218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09192025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28148996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09107181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09428264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09169985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28149036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09004208 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548977 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509561 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544814 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382372 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10544813 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382374 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382375 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548976 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509560 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10382369 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10544812 | - | - |
| Mouse Genome 430 2.0 Array | 1433472_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1433472_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100017515.mm.1 | NM_001048057.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017515.mm.1 | NM_001048057.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017515.mm.1 | NM_001048058.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017515.mm.1 | NM_001048058.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017515.mm.1 | NM_023372.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017515.mm.1 | NM_023372.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017515.mm.1 | TC1100001874.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1433472_x_at | - | - |
| Mouse Expression Set 430 | 1433472_PM_x_at | - | - |
| Murine Genome U74v2 Set | 99626_PM_i_at | - | - |
| Murine Genome U74v2 Set | 99627_r_at | - | - |
| Murine Genome U74v2 Set | 99627_PM_r_at | - | - |
| Murine Genome U74v2 Set | 99626_i_at | - | - |
| Murine Genome U74v2 Set | 169427_i_at | - | - |
| Murine Genome U74v2 Set | 169427_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433472_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433472_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 898302 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 70040 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4060138 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 70040 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060138 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6200601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000070040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200601 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2594971 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223383 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2594971 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223383 | - | - |
| mouse-6 v1.1 (Illumina) | scl40751.1_9-S | - | - |
| mouse-6 v1.1 (Illumina) | scl067671.4_8-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0067671.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060138 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 70040 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6200601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067671.1_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40751.1_9-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067671.4_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060138 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 70040 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3135931 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2594971 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3135931 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2594971 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067671.4_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40751.1_9-S | - | - |

Rpl38
Genomic Map