Assay Details
| Gene Symbol: Hn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC055351.1 | - | 316 | 103 | ||
| Gene Symbol: Gm3373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040090.1 | - | 381 | 3987869 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC040090.1 transcripts.
Set Membership: |
|

Hn1l
Genomic Map