| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400000460.mm.1 |
NM_013526.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000460.mm.1 |
NM_013526.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000460.mm.1 |
TC0400000069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400000459.mm.1 |
NM_013526.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000459.mm.1 |
NM_013526.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000459.mm.1 |
TC0400000069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400000462.mm.1 |
NM_013526.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000462.mm.1 |
NM_013526.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000462.mm.1 |
TC0400000069.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400000461.mm.1 |
NM_013526.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000461.mm.1 |
NM_013526.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400000461.mm.1 |
TC0400000069.mm.1 |
MTA TC |