Assay Details
| Gene Symbol: Bmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007555.3 | NP_031581.2 | 1 | 883 | 120 | |
| GenBank mRNA | AK033362.1 | - | 1 | 886 | 120 | |
| BC100751.1 | - | 1 | 154 | 120 | ||
| BC100752.1 | - | 1 | 154 | 120 | ||
| BC100753.1 | - | 1 | 154 | 120 | ||
| BC100754.1 | - | 1 | 154 | 120 | ||
| BC141283.1 | - | 1 | 686 | 9055895 | 120 | |
| L41145.1 | - | 1 | 155 | 120 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bmp5
Genomic Map