Assay Details
| Gene Symbol: H2-T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008208.4 | NP_032234.3 | 6 | 1943 | 135 | |
| GenBank mRNA | AK033602.1 | - | 6 | 1944 | 135 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1956048 | NM_008208.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1956048 | AK033602.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409415 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126705 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118436 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1956048 | NM_008208.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1956048 | AK033602.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P129127 | AY989871.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P129127 | D86082.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P129127 | BC125380.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P129127 | BC125378.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P129127 | NM_008208.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31163002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03312706 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126705 | - | - |
| Whole Mouse Genome G4122A | A_51_P129127 | AY989871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P129127 | D86082.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P129127 | BC125380.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P129127 | BC125378.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P129127 | NM_008208.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P246277 | M16810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P246277 | M16810.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109497 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121577 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126705 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13421271 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13112355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31163011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13423553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13201778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13455771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13455756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13546941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13546938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13421270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13505289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13546934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13067538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31163010 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450743 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450740 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450742 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450738 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450741 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450739 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450736 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450735 | - | - |
| Mouse Genome 430 2.0 Array | 1460418_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452548_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452548_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427726_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460418_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452547_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1427726_at | - | - |
| Mouse Genome 430 2.0 Array | 1452547_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15043_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15043_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700019250.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019250.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019250.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019255.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019255.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019255.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019260.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019260.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019260.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019254.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019254.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019254.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019256.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019256.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019256.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019259.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019259.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019259.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019264.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019264.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019264.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019249.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019249.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019249.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019266.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019266.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019266.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019246.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019246.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019246.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019248.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019248.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019248.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019263.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019263.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019263.mm.1 | TC1700002006.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019251.mm.1 | NM_008208.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019251.mm.1 | NM_008208.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019251.mm.1 | TC1700002006.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460418_x_at | - | - |
| Mouse Expression Set 430 | 1452548_PM_x_at | - | - |
| Mouse Expression Set 430 | 1452547_s_at | - | - |
| Mouse Expression Set 430 | 1460418_PM_x_at | - | - |
| Mouse Expression Set 430 | 1452548_x_at | - | - |
| Mouse Expression Set 430 | 1427726_PM_at | - | - |
| Mouse Expression Set 430 | 1452547_PM_s_at | - | - |
| Mouse Expression Set 430 | 1427726_at | - | - |
| Murine Genome U74v2 Set | 102692_PM_s_at | - | - |
| Murine Genome U74v2 Set | 98284_f_at | - | - |
| Murine Genome U74v2 Set | 96976_PM_f_at | - | - |
| Murine Genome U74v2 Set | 98284_PM_f_at | - | - |
| Murine Genome U74v2 Set | 96976_f_at | - | - |
| Murine Genome U74v2 Set | 102692_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460418_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452548_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452548_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452547_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427726_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452547_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460418_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427726_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2190047 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2190047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002190047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259837 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259837 | - | - |
| mouse-6 v1.1 (Illumina) | scl015043.1_307-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2190047 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl015043.1_307-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1259837 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259837 | - | - |

H2-T3
Genomic Map