Assay Details
| Gene Symbol: Rpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025592.3 | NP_079868.1 | 4 | 366 | 109 | |
| GenBank mRNA | AK010639.2 | - | 4 | 367 | 109 | |
| AK012335.1 | - | 4 | 367 | 109 | ||
| AK151707.1 | - | 4 | 361 | 109 | ||
| AK169650.1 | - | 4 | 367 | 109 | ||
| BC025884.1 | - | 4 | 348 | 109 | ||
| BC085091.1 | - | 2 | 1055 | 109 | ||
| BC094634.1 | - | 4 | 350 | 1333082 | 109 | |
| BC125652.1 | - | 4 | 348 | 40130167 | 109 | |
| BC125656.1 | - | 4 | 348 | 40130171 | 109 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151630 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151628 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151631 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149020 | AK010639.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149020 | NM_025592.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149020 | BC025884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149020 | BC085091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2149020 | BC094634.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10953 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140102 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149020 | AK010639.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149020 | NM_025592.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149020 | BC025884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149020 | BC085091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2149020 | BC094634.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132715 | AK010639.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132715 | AK012335.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132715 | NM_025592.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132715 | AK151707.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P132715 | AK169650.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151624 | - | - |
| Whole Mouse Genome G4122A | A_51_P132715 | AK010639.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132715 | AK012335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132715 | NM_025592.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132715 | AK151707.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P132715 | AK169650.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01234537 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01670538 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01234530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01670540 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151628 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151631 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01234530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01340912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01593485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01234539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01670539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02009613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01670538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01234543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01234537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01234540 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10482269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482272 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10350817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10457732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482268 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436840_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455950_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454856_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454833_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434231_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10350816 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10457731 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10482267 | - | - |
| Mouse Genome 430 2.0 Array | 1454856_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1454833_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416243_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455950_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436840_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436840_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455950_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1434231_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1454833_at | - | - |
| Mouse Genome 430 2.0 Array | 1454856_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1416243_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434231_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200029408.mm.1 | NM_025592.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029408.mm.1 | NM_025592.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029408.mm.1 | TC0200003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200029404.mm.1 | NM_025592.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029404.mm.1 | NM_025592.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029404.mm.1 | TC0200003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200029406.mm.1 | NM_025592.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029406.mm.1 | NM_025592.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029406.mm.1 | TC0200003433.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200029405.mm.1 | NM_025592.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029405.mm.1 | NM_025592.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029405.mm.1 | TC0200003433.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454856_PM_x_at | - | - |
| Mouse Expression Set 430 | 1454833_PM_at | - | - |
| Mouse Expression Set 430 | 1436840_PM_x_at | - | - |
| Mouse Expression Set 430 | 1416243_PM_a_at | - | - |
| Mouse Expression Set 430 | 1455950_PM_x_at | - | - |
| Mouse Expression Set 430 | 1416243_a_at | - | - |
| Mouse Expression Set 430 | 1454833_at | - | - |
| Mouse Expression Set 430 | 1436840_x_at | - | - |
| Mouse Expression Set 430 | 1455950_x_at | - | - |
| Mouse Expression Set 430 | 1434231_PM_x_at | - | - |
| Mouse Expression Set 430 | 1454856_x_at | - | - |
| Mouse Expression Set 430 | 1434231_x_at | - | - |
| Murine Genome U74v2 Set | 93098_at | - | - |
| Murine Genome U74v2 Set | 93098_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454833_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434231_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416243_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436840_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436840_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454856_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434231_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455950_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454856_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454833_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455950_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416243_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454833_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454833_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 894074 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6940070 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940070 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227440 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227440 | - | - |
| mouse-6 v1.1 (Illumina) | scl066489.3_56-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940070 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066489.3_56-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940070 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1227440 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227440 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066489.3_56-S | - | - |

Rpl35
Genomic Map