Assay Details
| Gene Symbol: Serpinb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198680.2 | NP_941373.1 | 2 | 59 | 80 | |
| GenBank mRNA | AK003220.1 | - | 2 | 60 | 80 | |
| BC099584.1 | - | 2 | 56 | 30291007 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003220.1, BC099584.1, NM_198680.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115152 | AY367774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115152 | BC099584.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115152 | NM_198680.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115152 | AK003220.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P115152 | AY144685.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50954 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115152 | AY367774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115152 | BC099584.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115152 | NM_198680.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115152 | AK003220.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P115152 | AY144685.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20511842 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04217519 | - | - |
| Whole Mouse Genome G4122A | A_51_P207763 | BC099584.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P207763 | NM_198680.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P207763 | AK003220.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115152 | AY367774.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115152 | BC099584.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115152 | NM_198680.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115152 | AK003220.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115152 | AY144685.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00549584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20511775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20511795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00814197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20511800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00016303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00814173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00814191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00814174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00549584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20511802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00016305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10357089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357086 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10357084 | - | - |
| Mouse Genome 430 2.0 Array | 1422940_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1422939_at | - | - |
| Mouse Genome 430 2.0 Array | 1422940_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1422939_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100023504.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023504.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023504.mm.1 | TC0100003884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023506.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023506.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023506.mm.1 | TC0100003884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023502.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023502.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023502.mm.1 | TC0100003884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023508.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023508.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023508.mm.1 | TC0100003884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023505.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023505.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023505.mm.1 | TC0100003884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023501.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023501.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023501.mm.1 | TC0100003884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023503.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023503.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023503.mm.1 | TC0100003884.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023507.mm.1 | NM_198680.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023507.mm.1 | NM_198680.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023507.mm.1 | TC0100003884.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422939_PM_at | - | - |
| Mouse Expression Set 430 | 1422940_x_at | - | - |
| Mouse Expression Set 430 | 1422940_PM_x_at | - | - |
| Mouse Expression Set 430 | 1422939_at | - | - |
| Murine Genome U74v2 Set | 170908_f_at | - | - |
| Murine Genome U74v2 Set | 171516_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170906_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170906_r_at | - | - |
| Murine Genome U74v2 Set | 171516_r_at | - | - |
| Murine Genome U74v2 Set | 170908_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422940_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422939_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422940_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422939_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 543245 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3440093 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370041 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370041 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2669321 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2630163 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2630163 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2669321 | - | - |
| mouse-6 v1.1 (Illumina) | scl0383548.1_52-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00383548.1_294-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440093 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370041 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00383548.1_294-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0383548.1_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440093 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370041 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2630163 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3001787 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2669321 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2669321 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2630163 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3001787 | - | - |

Serpinb3b
Genomic Map