Assay Details
| Gene Symbol: Prps1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037746.3 | NP_001032835.2 | 2 | 1544 | 138 | |
| GenBank mRNA | AK134500.1 | - | 2 | 1545 | 138 | |
| BC110357.1 | - | 2 | 1447 | 5293637 | 138 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461535 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461538 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461536 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461537 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461539 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975670 | AK134500.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975670 | BC110357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975670 | NM_001037746.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895623 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169472 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975670 | AK134500.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975670 | BC110357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1975670 | NM_001037746.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28461524 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169472 | - | - |
| Whole Mouse Genome G4122A | A_52_P604025 | AW552637.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P604025 | AK032842.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P604025 | AK089022.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P604025 | NM_001037746.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P644248 | AK037944.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461537 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461536 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461539 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461538 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461535 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28461534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28461509 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10109762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10109763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09842646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10217048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10299110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P09999384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10217046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28461562 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10395858 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395856 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10395857 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435671_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433903_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454814_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10395855 | - | - |
| Mouse Genome 430 2.0 Array | 1454814_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1433903_at | - | - |
| Mouse Genome 430 2.0 Array | 1433903_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454814_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 328099_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 328099_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200003336.mm.1 | NM_001037746.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003336.mm.1 | NM_001037746.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003336.mm.1 | TC1200000486.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003339.mm.1 | NM_001037746.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003339.mm.1 | NM_001037746.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003339.mm.1 | TC1200000486.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200003337.mm.1 | NM_001037746.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003337.mm.1 | NM_001037746.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200003337.mm.1 | TC1200000486.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454814_PM_s_at | - | - |
| Mouse Expression Set 430 | 1433903_at | - | - |
| Mouse Expression Set 430 | 1454814_s_at | - | - |
| Mouse Expression Set 430 | 1433903_PM_at | - | - |
| Mouse Expression Set 430 | 1448192_PM_s_at | - | - |
| Mouse Expression Set 430 | 1448192_s_at | - | - |
| Murine Genome U74v2 Set | 104039_at | - | - |
| Murine Genome U74v2 Set | 161897_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161897_f_at | - | - |
| Murine Genome U74v2 Set | 104039_PM_at | - | - |
| Murine Genome U74v2 Set | 95018_r_at | - | - |
| Murine Genome U74v2 Set | 95018_PM_r_at | - | - |
| Murine Genome U74v2 Set | 168480_PM_at | - | - |
| Murine Genome U74v2 Set | 168822_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168480_at | - | - |
| Murine Genome U74v2 Set | 168822_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454814_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433903_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448192_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1433903_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448192_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1454814_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454814_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1454814_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433903_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1433903_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160212 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160212 | - | - |
| mouse-6 v1.1 (Illumina) | GI_85702277-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160212 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160212 | - | - |

Prps1l3
Genomic Map