Assay Details
| Gene Symbol: Lsmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BY080835.1 | - | 2 - 3 | 197 | 65 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117950 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26809774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07562979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07508016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07508022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809816 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07562969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07562961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26809814 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10596677 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596678 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596676 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596679 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10596675 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_3030685 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3030685 | - | - |

Lsmem2
Genomic Map