Assay Details
| Gene Symbol: Ces1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081372.1 | NP_001074841.1 | 8 - 9 | 1044 | 63 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P282905 | NM_001081372.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P282905 | NM_001081372.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661051 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574150 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49269 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83761 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P282905 | NM_001081372.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P282905 | NM_001081372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180988 | - | - |
| Whole Mouse Genome G4122A | A_52_P282905 | NM_001081372.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P282905 | NM_001081372.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26069722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06756257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06932108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07276049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07276034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07276035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07275990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26069731 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10580623 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10580622 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_3112143 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3112143 | - | - |

Ces1b
Genomic Map