Assay Details
| Gene Symbol: Nacad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081652.1 | NP_001075121.1 | 3 - 4 | 4321 | 77 | |
| GenBank mRNA | AB182283.1 | - | 3 - 4 | 3838 | 77 | |
| BC072589.1 | - | 3 - 4 | 4321 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nacad
Genomic Map