Assay Details
Gene Symbol: Nacad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081652.1 | NP_001075121.1 | 5 - 6 | 4541 | 74 | |
GenBank mRNA | AB182283.1 | - | 5 - 6 | 4058 | 74 | |
BC072589.1 | - | 5 - 6 | 4541 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
