Assay Details
| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004173.2 | NP_001004173.1 | 2 - 3 | 321 | 100 | |
| GenBank mRNA | BC049995.1 | - | 2 - 3 | 322 | 6399201 | 100 |
| BC147843.1 | - | 2 - 3 | 510 | 9087560 | 100 | |
| BC147850.1 | - | 2 - 3 | 510 | 9087567 | 100 | |
| BY297240.1 | - | 2 - 3 | 320 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sgpp2
Genomic Map