Assay Details
| Gene Symbol: Wfdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080550.2 | NP_001074019.1 | 5 - 6 | 940 | 93 | |
| NM_001276431.1 | NP_001263360.1 | 5 - 6 | 940 | 93 | ||
| NM_001276432.1 | NP_001263361.1 | 5 - 6 | 1769 | 93 | ||
| GenBank mRNA | AK136599.1 | - | 5 - 6 | 1770 | 93 | |
| AY669506.1 | - | 5 - 6 | 940 | 93 | ||
| AY757956.1 | - | 5 - 6 | 940 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146768 | AY757956.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2146768 | NM_001080550.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042570 | AK136599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042577 | AK136599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042577 | AY669506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042577 | AY757956.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042577 | NM_001080550.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768760 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275044 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315568 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21447 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146768 | AY757956.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2146768 | NM_001080550.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042577 | AK136599.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042577 | AY669506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042577 | AY757956.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042577 | NM_001080550.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042570 | AK136599.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04772524 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04772537 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00550122 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00550130 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04772537 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00550122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21818758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01691354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01741969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02038005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02172172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01243528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01338623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02038004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01315078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01770937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01691357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01305735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01691351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01495629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02065525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01782058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21818852 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489514 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489520 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489512 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200043677.mm.1 | NM_001080550.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043677.mm.1 | NM_001276432.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043677.mm.1 | TC0200005122.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043679.mm.1 | NM_001080550.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043679.mm.1 | NM_001276431.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043679.mm.1 | NM_001276432.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043679.mm.1 | TC0200005122.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043676.mm.1 | NM_001080550.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043676.mm.1 | NM_001276431.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043676.mm.1 | NM_001276432.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043676.mm.1 | TC0200005122.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043682.mm.1 | NM_001080550.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043682.mm.1 | NM_001276431.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043682.mm.1 | NM_001276432.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043682.mm.1 | TC0200005122.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043678.mm.1 | NM_001080550.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043678.mm.1 | NM_001276432.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043678.mm.1 | TC0200005122.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200043681.mm.1 | NM_001080550.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043681.mm.1 | TC0200005122.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223633 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2838051 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2838051 | - | - |

Wfdc8
Genomic Map