Assay Details
| Gene Symbol: Larp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024526.2 | NP_001019697.2 | 10 - 11 | 1295 | 112 | |
| NM_001080948.2 | NP_001074417.1 | 10 - 11 | 1308 | 112 | ||
| NM_001284521.1 | NP_001271450.1 | 10 - 11 | 1282 | 112 | ||
| NM_001284522.1 | NP_001271451.1 | 10 - 11 | 1305 | 112 | ||
| NM_001284523.1 | NP_001271452.1 | 10 - 11 | 1298 | 112 | ||
| GenBank mRNA | AK049756.1 | - | 10 - 11 | 1222 | 112 | |
| AK139636.1 | - | 10 - 11 | 1303 | 112 | ||
| AK139909.1 | - | 10 - 11 | 1146 | 112 | ||
| AK165392.1 | - | 10 - 11 | 1243 | 112 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp4
Genomic Map