Assay Details
| Gene Symbol: Aurkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080965.1 | NP_001074434.1 | 5 - 6 | 678 | 95 | |
| NM_001080966.1 | NP_001074435.1 | 5 - 6 | 964 | 95 | ||
| NM_020572.2 | NP_065597.2 | 5 - 6 | 842 | 95 | ||
| GenBank mRNA | AF054620.1 | - | 5 - 6 | 929 | 95 | |
| BC064780.1 | - | 5 - 6 | 846 | 30476803 | 95 | |
| BC100337.1 | - | 5 - 6 | 599 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cell cycle
mitotic spindle organization
mitotic nuclear division
regulation of signal transduction
phosphorylation
regulation of cytokinesis
histone-serine phosphorylation
cell division
meiotic cell cycle
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
histone serine kinase activity
protein phosphorylation
cell cycle
mitotic spindle organization
mitotic nuclear division
regulation of signal transduction
phosphorylation
regulation of cytokinesis
histone-serine phosphorylation
cell division
meiotic cell cycle
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
histone serine kinase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999511 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P131931 | AF054620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P131931 | AF054620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999511 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512293 | AA545317.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P512293 | AF054620.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06123511 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06123506 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06123564 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06123550 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01478695 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06123591 | - | - |
| Whole Mouse Genome G4122A | A_52_P465718 | BC064780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P465718 | BC100337.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P465718 | BG080448.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P465718 | DV648982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P465718 | NM_001080965.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512293 | AA545317.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P512293 | AF054620.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102050 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131931 | AF054620.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109295 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129507 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06123550 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06123506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24957730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05895631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06384095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06416979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06278154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06278140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05895622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06278150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06278134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05895626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06384094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24957761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06278145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559993 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512681 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549898 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559995 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549890 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449364_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559989 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512680 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560004 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549889 | - | - |
| Mouse Genome 430 2.0 Array | 1449364_at | - | - |
| Mouse Genome 430 2.0 Array | 1449364_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000956.mm.1 | TC0700000099.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000956.mm.1 | NM_001080965.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000956.mm.1 | NM_001080965.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000956.mm.1 | NM_001080966.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000956.mm.1 | NM_001080966.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000956.mm.1 | NM_020572.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000956.mm.1 | NM_020572.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000954.mm.1 | TC0700000099.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000954.mm.1 | NM_001080965.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000954.mm.1 | NM_001080965.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000954.mm.1 | NM_001080966.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000954.mm.1 | NM_001080966.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000954.mm.1 | NM_020572.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000954.mm.1 | NM_020572.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000955.mm.1 | TC0700000099.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000955.mm.1 | NM_001080965.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000955.mm.1 | NM_001080965.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000955.mm.1 | NM_001080966.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000955.mm.1 | NM_001080966.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000955.mm.1 | NM_020572.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000955.mm.1 | NM_020572.2 | Refseq |
| Mouse Expression Set 430 | 1449364_at | - | - |
| Mouse Expression Set 430 | 1449364_PM_at | - | - |
| Murine Genome U74v2 Set | 129054_PM_i_at | - | - |
| Murine Genome U74v2 Set | 129054_i_at | - | - |
| Murine Genome U74v2 Set | 170449_at | - | - |
| Murine Genome U74v2 Set | 170449_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449364_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449364_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449364_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449364_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 595523 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4060687 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060687 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060687 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2725246 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2725246 | - | - |
| mouse-6 v1.1 (Illumina) | scl0020871.1_53-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060687 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020871.1_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060687 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3136523 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2725246 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3136523 | - | - |

Aurkc
Genomic Map