Assay Details
Gene Symbol: Hist1h2ak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178183.2 | NP_835490.1 | 1 | 431 | 73 | |
GenBank mRNA | BC119335.1 | - | 1 | 431 | 8734084 | 73 |
Gene Symbol: Hist1h2ai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178182.2 | NP_835489.1 | 1 | 356 | 73 | |
GenBank mRNA | BC099406.1 | - | 356 | 30789778 | 73 | |
BC116373.2 | - | 1 | 346 | 40100893 | 73 | |
BC127164.1 | - | 1 | 346 | 40100895 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874436 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874434 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084652 | NM_178183.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2084652 | BC119335.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498208 | NM_178183.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498208 | BC119335.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498208 | NM_178183.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498208 | BC119335.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498208 | NM_178183.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498208 | BC119335.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498208 | NM_178183.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498208 | BC119335.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084652 | NM_178183.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2084652 | BC119335.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874435 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874436 | - | - |
Whole Mouse Genome G4122A | A_52_P498208 | NM_178183.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498208 | BC119335.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498208 | NM_178183.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498208 | BC119335.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P651235 | BC099406.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P651235 | NM_178182.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128873 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10893034 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874434 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874213 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408080 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404047 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408120 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404047 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403939 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408096 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408073 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500331 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408086 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408096 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404027 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408120 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10403940 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408078 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10409221 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408112 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408073 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408079 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404027 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10494393 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408086 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408112 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404045 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408118 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408094 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404026 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408085 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403955 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408111 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403955 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408085 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403938 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408094 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404045 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404026 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10403938 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408111 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408077 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408118 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408072 | - | - |
Affymetrix Mouse Clariom S Array | PSR1300009835.mm.1 | TC1300001558.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001166.mm.1 | TC1300000173.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300009836.mm.1 | NM_178183.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300009836.mm.1 | TC1300001558.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300009837.mm.1 | TC1300001558.mm.1 | MTA TC |
Mouse Expression Set 430 | 1438009_at | - | - |
Mouse Expression Set 430 | 1418366_at | - | - |
Mouse Expression Set 430 | 1418367_PM_x_at | - | - |
Mouse Expression Set 430 | 1418367_x_at | - | - |
Mouse Expression Set 430 | 1418366_PM_at | - | - |
Mouse Expression Set 430 | 1438009_PM_at | - | - |
Murine Genome U74v2 Set | 94805_f_at | - | - |
Murine Genome U74v2 Set | 94805_f_at | - | - |
Murine Genome U74v2 Set | 94805_PM_f_at | - | - |
Murine Genome U74v2 Set | 94805_PM_f_at | - | - |
Murine Genome U74v2 Set | 167053_PM_at | - | - |
Murine Genome U74v2 Set | 167053_at | - | - |
Murine Genome U74v2 Set | 167053_at | - | - |
Murine Genome U74v2 Set | 167053_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1438009_at | - | - |
Mouse Genome 430A 2.0 Array | 1418367_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438009_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418366_at | - | - |
Mouse Genome 430A 2.0 Array | 1418366_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418367_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 893035 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102480368 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4540039 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102570446 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004540039 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240660 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1238276 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240660 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1245449 | - | - |
mouse-6 v1.1 (Illumina) | GI_30061392-S | - | - |
mouse-6 v1.1 (Illumina) | scl0319191.1_321-S | - | - |
mouse-6 v1.1 (Illumina) | GI_30061370-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4540039 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102480368 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102570446 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0319191.1_321-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102570446 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102480368 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540039 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2847144 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2847144 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2745212 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238276 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0319191.1_321-S | - | - |
