Assay Details
| Gene Symbol: Rab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293652.1 | NP_001280581.1 | 6 - 7 | 823 | 80 | |
| NM_001293653.1 | NP_001280582.1 | 4 - 5 | 484 | 80 | ||
| NM_001293654.1 | NP_001280583.1 | 5 - 6 | 650 | 80 | ||
| NM_001293655.1 | NP_001280584.1 | 4 - 5 | 563 | 80 | ||
| NM_009005.3 | NP_033031.2 | 4 - 5 | 601 | 80 | ||
| GenBank mRNA | AB232599.1 | - | 3 - 4 | 395 | 80 | |
| AK005004.1 | - | 4 - 5 | 563 | 80 | ||
| AK057671.1 | - | 4 - 5 | 530 | 80 | ||
| AK139914.1 | - | 4 - 5 | 554 | 80 | ||
| AK148117.1 | - | 4 - 5 | 594 | 80 | ||
| AK151078.1 | - | 4 - 5 | 575 | 80 | ||
| AK152666.1 | - | 6 - 7 | 798 | 80 | ||
| AK154234.1 | - | 4 - 5 | 582 | 80 | ||
| AK158971.1 | - | 4 - 5 | 485 | 80 | ||
| AK159507.1 | - | 4 - 5 | 530 | 80 | ||
| AK159928.1 | - | 4 - 5 | 502 | 80 | ||
| AK183086.1 | - | 1 - 2 | 59 | 80 | ||
| BC004597.1 | - | 4 - 5 | 519 | 3584891 | 80 | |
| BC086793.1 | - | 4 - 5 | 517 | 6414012 | 80 | |
| CT010352.1 | - | 3 - 4 | 395 | 80 | ||
| X89650.1 | - | 4 - 5 | 551 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab7
Genomic Map