Assay Details
| Gene Symbol: Cd68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291058.1 | NP_001277987.1 | 5 - 6 | 956 | 109 | |
| NR_110993.1 | 4 - 5 | 522 | 109 | |||
| GenBank mRNA | AK002264.1 | - | 5 - 6 | 914 | 109 | |
| AK170443.1 | - | 5 - 6 | 880 | 109 | ||
| AW123877.1 | - | 1 - 2 | 109 | |||
| BC021637.1 | - | 5 - 6 | 897 | 3981024 | 109 | |
| X68273.1 | - | 5 - 6 | 939 | 109 | ||
More Information
Set Membership: |
|

Cd68
Genomic Map