Assay Details
Gene Symbol: Olfr618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147047.2 | NP_667258.2 | 1 | 867 | 130 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2144160 | NM_147047.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416759 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147669 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2144160 | NM_147047.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P292859 | NM_147047.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147669 | - | - |
Whole Mouse Genome G4122A | A_51_P292859 | NM_147047.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135984 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25384130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384064 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06283298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384036 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06335102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06195264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06093648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06093650 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06221875 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384042 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25384069 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10555804 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10555803 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2630273 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2861686 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2861686 | - | - |
