Assay Details
| Gene Symbol: Pcdhac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003672.1 | NP_001003672.1 | 1 - 2 | 2563 | 102 | |
| GenBank mRNA | AY013769.1 | - | 1 - 2 | 2563 | 102 | |
More Information
Set Membership: |
|

Pcdhac2
Genomic Map