Assay Details
| Gene Symbol: Akr1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009658.3 | NP_033788.3 | 2 - 3 | 317 | 86 | |
| GenBank mRNA | AK028079.1 | - | 2 - 3 | 272 | 86 | |
| AK138000.1 | - | 2 - 3 | 270 | 86 | ||
| AK139250.1 | - | 2 - 3 | 272 | 86 | ||
| AK145392.1 | - | 2 - 3 | 273 | 86 | ||
| AK149861.1 | - | 2 - 3 | 307 | 86 | ||
| AK150197.1 | - | 2 - 3 | 272 | 86 | ||
| AK153949.1 | - | 2 - 3 | 301 | 86 | ||
| AK164682.1 | - | 2 - 3 | 272 | 86 | ||
| AK170663.1 | - | 2 - 3 | 319 | 86 | ||
| BC004725.1 | - | 2 - 3 | 264 | 3582788 | 86 | |
| BC021655.1 | - | 2 - 3 | 310 | 3708927 | 86 | |
| BC085310.1 | - | 2 - 3 | 249 | 6485416 | 86 | |
| BY749711.1 | - | 2 - 3 | 317 | 86 | ||
| D32250.1 | - | 2 - 3 | 258 | 86 | ||
| L39795.1 | - | 2 - 3 | 249 | 86 | ||
| U29152.1 | - | 2 - 3 | 268 | 86 | ||
| Gene Symbol: Gm6644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037965.1 | 1 | 272 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Akr1b3
Genomic Map