| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100011039.mm.1 |
NM_001159730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011039.mm.1 |
NM_001159730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011039.mm.1 |
NM_024458.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011039.mm.1 |
NM_024458.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011039.mm.1 |
TC0100001339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011040.mm.1 |
NM_001159730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011040.mm.1 |
NM_001159730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011040.mm.1 |
NM_024458.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011040.mm.1 |
NM_024458.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011040.mm.1 |
TC0100001339.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100011037.mm.1 |
NM_001159730.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011037.mm.1 |
NM_001159730.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011037.mm.1 |
NM_024458.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011037.mm.1 |
NM_024458.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100011037.mm.1 |
TC0100001339.mm.1 |
MTA TC |