Assay Details
| Gene Symbol: Ddx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025860.3 | NP_080136.2 | 5 - 6 | 829 | 86 | |
| GenBank mRNA | AK019845.1 | - | 5 - 6 | 753 | 86 | |
| AK075864.1 | - | 5 - 6 | 766 | 86 | ||
| BC017160.1 | - | 5 - 6 | 749 | 86 | ||
| BC028246.1 | - | 5 - 6 | 800 | 2582151 | 86 | |
| BC103776.1 | - | 5 - 6 | 766 | 6492142 | 86 | |
| BC141230.1 | - | 5 - 6 | 768 | 9055842 | 86 | |
| BC141231.1 | - | 5 - 6 | 768 | 9055843 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ddx18
Genomic Map