Assay Details
| Gene Symbol: Fcamr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170632.1 | NP_001164103.1 | 3 - 4 | 456 | 128 | |
| NM_144960.2 | NP_659209.2 | 3 - 4 | 288 | 128 | ||
| GenBank mRNA | AB048834.1 | - | 3 - 4 | 300 | 128 | |
| BC110549.1 | - | 3 - 4 | 459 | 40047147 | 128 | |
| BC110550.1 | - | 3 - 4 | 291 | 40047153 | 128 | |
| FJ011703.1 | - | 2 - 3 | 369 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcamr
Genomic Map